Learn More
Kingfisher Biotech Inc Feline CCL7 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1854F025
Encompass
The Feline CCL7 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline CCL7 applications are for cell culture, ELISA standard, and Western Blot Control. Feline CCL7 yeast-derived recombinant protein can be purchased in multiple sizes. Feline CCL7 Specifications: (Molecular Weight: 9.0 kDa) (Amino Acid Sequence: QPEGINSPTTCCYRFTVRKIPIQKLKSYRRITSSYCPREAVIFKTKQAKELCADPKQKWVQDFMEYLDKKTQTSKL (76)) (Gene ID: 101096931). For research use only. Made in the USA
Catalog No. 50-253-2693