Learn More
Raybiotech Inc G-Series Feline Cytokine Array 1 Kit.
Catalog No. 501950350
Encompass_Preferred
Semi-quantitative measurement of 10 Feline cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Fas (TNFRSF6/Apo-1), IFN-gamma, IL-1 beta (IL-1 F2), IL-2, IL-4, IL-5, IL-8 (CXCL8), IL-10, IL-12 p40, RANTES (CCL5)
Catalog No. 50-195-0350 Supplier Raybiotech Inc Supplier No. GSFCYT11
Semi-quantitative measurement of 10 Feline cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Fas (TNFRSF6/Apo-1), IFN-gamma, IL-1 beta (IL-1 F2), IL-2, IL-4, IL-5, IL-8 (CXCL8), IL-10, IL-12 p40, RANTES (CCL5)