Learn More
Med Vet International Feline Declaw Forceps, Stainless Steel, German, 4"
Catalog No. 5000584002
Encompass_Preferred
These forceps are very useful in feline declaws especially when doing the procedure with a laser. The wide end grabs the claw firmly so a precise incision can be made.
Catalog No. 50-005-84002 Supplier Med Vet International Supplier No. XJOR1026
These forceps are very useful in feline declaws especially when doing the procedure with a laser. The wide end grabs the claw firmly so a precise incision can be made.