Learn More
Med Vet International Feline Dental Extractor, 6”, J0042T, Each
Supplier: Med Vet International JOR42T
This extractor is smaller and lighter weight with special jaws that close together along their whole interface. Especially designed for small dog and feline teeth or tooth fragments. Also useful in removing tartar/calculus.