Learn More
Kingfisher Biotech Inc Feline Erythropoietin Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Catalog No. 502533205
Encompass
The Feline Erythropoietin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline Erythropoietin applications are for cell culture, ELISA standard, and Western Blot Control. Feline Erythropoietin yeast-derived recombinant protein can be purchased in multiple sizes. Feline Erythropoietin Specifications: (Molecular Weight: 18.3 kDa) (Amino Acid Sequence: APPRLICDSR VLERYILGAR EAENVTMGCA EGCSFSENIT VPDTKVNFYT WKRMDVGQQA VEVWQGLALL SEAILRGQAL LANSSQPSET LQLHVDKAVS SLRSLTSLLR ALGAQKEATS LPEATSAAPL RTFTVDTLCK LFRIYSNFLR GKLTLYTGEA CRRGDR (166)) (Gene ID: 493801). For research use only. Made in the USA
Catalog No. 50-253-3205 Supplier Kingfisher Biotech Inc Supplier No. RP1524F100
The Feline Erythropoietin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline Erythropoietin applications are for cell culture, ELISA standard, and Western Blot Control. Feline Erythropoietin yeast-derived recombinant protein can be purchased in multiple sizes. Feline Erythropoietin Specifications: (Molecular Weight: 18.3 kDa) (Amino Acid Sequence: APPRLICDSR VLERYILGAR EAENVTMGCA EGCSFSENIT VPDTKVNFYT WKRMDVGQQA VEVWQGLALL SEAILRGQAL LANSSQPSET LQLHVDKAVS SLRSLTSLLR ALGAQKEATS LPEATSAAPL RTFTVDTLCK LFRIYSNFLR GKLTLYTGEA CRRGDR (166)) (Gene ID: 493801). For research use only. Made in the USA