Learn More
Kingfisher Biotech Inc Feline IL-1 beta Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0098F025
Encompass
The Feline IL-1 beta yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline IL-1 beta applications are for cell culture, ELISA standard, and Western Blot Control. The Feline IL-1 beta yeast-derived recombinant protein can be purchased in multiple sizes. Feline IL-1 beta Specifications: (Molecular Weight: 17.4 kDa) (Amino Acid Sequence: AAIQSQDYTFRDISQKSLVLSGSYELRALHLNGQNMNQQVVFRMSFVHGEENSKKIPVVLCIKKNNLYLSCVMKDGKPTLQLEMLDPKVYPKKKMEKRFVFNKTEIKGNVEFESSQFPNWYISTSQAEEMPVFLGNTKGGQDITDFIMESAS) (Gene ID: 768274). For research use only. Made in the USA
Catalog No. 50-241-3539