Learn More
Raybiotech Inc Feline IL-1 beta (IL-1 F2) ELISA Kit
Supplier:  Raybiotech Inc ELFIL1B1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 16 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects feline IL-1 beta. Other species not determined.
Catalog No. 50-194-8919