Learn More
Raybiotech Inc Feline IL-10 ELISA Kit
Supplier:  Raybiotech Inc ELFIL101
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects feline IL-10. Approximately 15% cross-reactivity with recombinant canine and 1% cross-reactivity with recombinant human IL-10 is observed.
Catalog No. 50-194-8917