Learn More
Kingfisher Biotech Inc Feline IL-12 p40 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP2079F025
Encompass
The Feline IL-12/IL-23 p40 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline IL-12/IL-23 p40 applications are for cell culture, ELISA standard, and Western Blot Control. Feline IL-12/IL-23 p40 yeast-derived recombinant protein can be purchased in multiple sizes. Feline IL-12/IL-23 p40 Specifications: (Molecular Weight: 35.0 kDa) (Amino Acid Sequence: IWELEKNVYV VELDWHPDAP GEMVVLTCNT PEEDDITWTS DQSSEVLGSG KTLTIQVKEF ADAGQYTCHK GGEVLSHSFL LIHKKEDGIW STDILREQKE SKNKIFLKCE AKNYSGRFTC WWLTAISTDL KFTVKSSRGS SDPQEVTCGA ATLSAEKVRV DNRDYKKYTV ECQEGSACPA AEESLPIEVV VDAIHKLKYE NYTSSFFIRD IIKPDPPKNL QLKPLKNSRH VEVSWEYPDT WSTPHSYFSL TFGVQVQGKN NREKKDRLSV DKTSAKVVCH KDAKIRVQAR DRYYSSSWSN WASVSCS (307)) (Gene ID: 768273). For research use only. Made in the USA
Catalog No. 50-253-2468