Learn More
Raybiotech Inc Feline IL-12 p40 ELISA Kit
Supplier:  Raybiotech Inc ELFIL12P401
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.16 ng/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: feline Fas, IFN-gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-8, IL-10, RANTES
Catalog No. 50-194-8918