Learn More
Kingfisher Biotech Inc Feline IL-13 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Catalog No. 502532495
Encompass
The Feline IL-13 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline IL-13 applications are for cell culture, ELISA standard, and Western Blot Control. Feline IL-13 yeast-derived recombinant protein can be purchased in multiple sizes. Feline IL-13 Specifications: (Molecular Weight: 12.7 kDa) (Amino Acid Sequence: SPGPHSRREL KELIEELVNI TQNQVSLCNG SMVWSVNLTT GMQYCAALES LINVSDCTAI QRTQRMLKAL CTQKPSAGQT ASERSRDTKI EVIQLVKNLL NHLRRNFRHG NFK (113)) (Gene ID: 101084678). For research use only. Made in the USA
Catalog No. 50-253-2495 Supplier Kingfisher Biotech Inc Supplier No. RP2053F005
The Feline IL-13 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline IL-13 applications are for cell culture, ELISA standard, and Western Blot Control. Feline IL-13 yeast-derived recombinant protein can be purchased in multiple sizes. Feline IL-13 Specifications: (Molecular Weight: 12.7 kDa) (Amino Acid Sequence: SPGPHSRREL KELIEELVNI TQNQVSLCNG SMVWSVNLTT GMQYCAALES LINVSDCTAI QRTQRMLKAL CTQKPSAGQT ASERSRDTKI EVIQLVKNLL NHLRRNFRHG NFK (113)) (Gene ID: 101084678). For research use only. Made in the USA