Learn More
Kingfisher Biotech Inc Feline IL-17A Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc KP1907F100
Encompass
The Feline IL-17A polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Feline IL-17A ELISA. The Feline IL-17A antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Bovine IL-17A - Weak) (Canine IL-17A - Moderate) (Caprine IL-17A - None) (Chicken IL-17A - None) (Cynomolgus Monkey IL-17A - None) (Dolphin IL-17A - Moderate) (Equine IL-17A - Weak) (Feline IL-17A - Strong) (Ferret IL-17A - None) (Guinea Pig IL-17A - None) (Human IL-17A - Weak) (Mallard IL-17A - None) (Mouse IL-17A - None) (Ovine IL-17A - Weak) (Rabbit IL-17A - Weak) (Rat IL-17A - Weak) (Swine IL-17A - Weak). For research use only. Made in the USA
Catalog No. 50-233-2087