Learn More
Kingfisher Biotech Inc Feline IL-17A Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1032F025
Encompass
The Feline IL-17A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline IL-17A applications are for cell culture, ELISA standard, and Western Blot Control. Feline IL-17A yeast-derived recombinant protein can be purchased in multiple sizes. Feline IL-17A Specifications: (Molecular Weight: 15.1 kDa) (Amino Acid Sequence: GIAFPQNPGC PTTEDKNFPQ HVKVNVNILN GNKSSRRPLD YYRRSTSPWS LHRNEDPERY PSVIWEAKCL HWGCVNTEGK EDHHMNSVPI QQEILVLRRE SRHCPHSFRL EKMLVTVGCT CVTPIVRHVV (130)) (Gene ID: 101095339). For research use only. Made in the USA
Catalog No. 50-241-3933