Learn More
Kingfisher Biotech Inc Rabbit VEGF-A 121 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1909F004
Encompass_Preferred
The Rabbit VEGF-A (121 aa) ELISA contains capture antibody, standard, and detection antibody for development of a Rabbit VEGF-A ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include VEGFA, MVCD1, VEGF, VPF, vascular endothelial growth factor A. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5590