Learn More
Raybiotech Inc Feline IL-2 ELISA Kit
Supplier:  Raybiotech Inc ELFIL21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 25 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects feline IL-2. Approximately 3% cross-reactivity with recombinant canine IL-2 is observed, approximately 1% cross reactivity with recombinant porcine IL-2 and recombinant human IL-2 is observed.
Catalog No. 50-194-8920