Learn More
Raybiotech Inc Feline IL-4 ELISA Kit
Supplier:  Raybiotech Inc ELFIL41
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.3 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit detects feline IL4. Less than 0.2% cross-reactivity with recombinant human IL-4, recombinant mouse IL-4, recombinant rat IL-4, recombinant cotton rat IL4, recombinant porcine IL-4, and recombinant canine IL4 is observed in this antibody pair.
Catalog No. 50-194-8921