Learn More
Kingfisher Biotech Inc Feline IL-5 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1152F025
Encompass
The Feline IL-5 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline IL-5 applications are for cell culture, ELISA standard, and Western Blot Control. Feline IL-5 yeast-derived recombinant protein can be purchased in multiple sizes. Feline IL-5 Specifications: (Molecular Weight: 13.2 kDa) (Amino Acid Sequence: IAVQSPMNRL VAETLALLST HRTLLIGDGN LMIPTPEHNN HQLCIEEVFQ GIDTLKNRTV PGDAVEKLFR NLSLIKEHID RQKKKCGGER WRVKKFLDYL QVFLGVINTE WTMES (115)) (Gene ID: 493803). For research use only. Made in the USA
Catalog No. 50-253-3609