Learn More
Raybiotech Inc Feline IL-5 ELISA Kit
Supplier:  Raybiotech Inc ELFIL51
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Feline IL-5.
Catalog No. 50-194-8922