Learn More
Raybiotech Inc Feline IL-8 (CXCL8) ELISA Kit
Supplier:  Raybiotech Inc ELFIL81
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 25 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects feline IL-8. Other species not determined
Catalog No. 50-194-8923