Learn More
Sino Biological Anti-IL-10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 67001T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Feline IL-10 (Catalog#67001-UNAE; P55029.2; Ser19-Ile178). IL-10 specific IgG was purified by Feline IL-10 affinity chromatography.
Catalog No. 50-241-0877