Learn More
Kingfisher Biotech Inc Feline M-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1702F025
Encompass
The Feline M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Feline M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Feline M-CSF Specifications: (Molecular Weight: 18.4 kDa) (Amino Acid Sequence: EEVSERCSHMIGNGHLQFLQQLIDSQMETSCQIAFEFVDQEQLKDPVCYLKKAFLLVQDIMEDTMRFKDNTPNANVIVTLQELSLRLSSCFTKDYEEQDKACVRTFHETPLQLLEKIKNVFNETKNLLKKDWNVFSKNCNKSFEKCSSQGHDRQQEGP (158)) (Gene ID: 101091467). For research use only. Made in the USA
Catalog No. 50-253-2921