Learn More
Raybiotech Inc Feline PDGF-BB ELISA Kit
Supplier:  Raybiotech Inc ELFPDGFB1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 5.4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Feline PDGF-BB.
Catalog No. 50-194-8924