Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Feline PDGF-BB ELISA Kit

Catalog No. EF9RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EF9RBX5 5 x 96 Tests
EF9RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EF9RBX5 Supplier Invitrogen™ Supplier No. EF9RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Feline PDGF-BB quantitates feline PDGF-BB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant feline PDGF-BB.

The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01127, P12919
Assay Range 5.49-4000 pg/mL
Assay Sensitivity 5.4 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human, Feline
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias becaplermin; c-sis; FLJ12858; H-PDGF-BB; IBGC5; PDGF; pdgf protein; PDGF subunit B; PDGF, B chain; PDGF2; PDGF-2; PDGFB; PDGF-B; platelet derived growth factor subunit B; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor B; platelet-derived growth factor B chain; platelet-derived growth factor beta; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); proto-oncogene c-Sis; Sis; SSVbecaplermin; c-sis; FLJ12858; H-PDGF-BB; IBGC5; PDGF; pdgf protein; PDGF subunit B; PDGF, B chain; PDGF2; PDGF-2; PDGFB; PDGF-B; platelet derived growth factor subunit B; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor B; platelet-derived growth factor B chain; platelet-derived growth factor beta; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); proto-oncogene c-Sis; Sis; SSV;
Gene ID (Entrez) 100135774, 5155
Gene Symbol PDGFB
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target PDGF-BB
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.