Learn More
Kingfisher Biotech Inc Feline SCF Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1302F100
Encompass
The Feline SCF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline SCF applications are for cell culture, ELISA standard, and Western Blot Control. Feline SCF yeast-derived recombinant protein can be purchased in multiple sizes. Feline SCF Specifications: (Molecular Weight: 18.4 kDa) (Amino Acid Sequence: GLCRNRVTDD VKDVTKLVAN LPKDYKIALK YVPGMDVLPS HCWISVMVEQ LSVSLTDLLD KFSNISEGLS NYSIIDKLVK IVDDLVECVE GHSSENVKKS SKSPEPRLFT PEEFFRIFNR SIDAFKDLEM VASKTSECVV SSTLSPEKDS RVSVTKPFML PPVAA (165)) (Gene ID: 493937). For research use only. Made in the USA
Catalog No. 50-253-3415