Learn More
Kingfisher Biotech Inc Feline SCF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0150F025
Encompass
The Feline SCF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline SCF applications are for cell culture, ELISA standard, and Western Blot Control. The Feline SCF yeast-derived recombinant protein can be purchased in multiple sizes. Feline SCF Specifications: (Molecular Weight: 27.9 kDa) (Amino Acid Sequence: GLCRNRVTDDVKDVTKLVANLPKDYKIALKYVPGMDVLPSHCWISVMVEQLSVSLTDLLDKFSNISEGLSNYSIIDKLVKIVDDLVECVEGHSSENVKKSSKSPEPRLFTPEEFFRIFNRSIDAFKDLEMVASKTSECVVSSTLSPEKDSRVSVTKPFMLPPVAASSLRNDSSSSNRKATNPIEDSSIQWAVMALPACFSLVIGFAFGAFYWKKKQPNLTRTVENIQINEEDNEISMLQEKEREFQEV (248)) (Gene ID: 493937). For research use only. Made in the USA
Catalog No. 50-241-3906