Learn More
Equitech Bio Inc Feline Serum Albumin Powder, ≥ 96% Purity, Refrigerated Storage, 100 mg
SDP
Catalog No. NC3640625
Encompass
EQUITECH BIO INC Feline Serum Albumin is a high-purity lyophilized powder ideal for various scientific applications. With ≥ 96% purity and preserved components such as fatty acids, it serves as a versatile serological diluent and molecular weight standard. Processed under cold, non-denaturing conditions, it maintains protein integrity, making it suitable for research and manufacturing purposes. Refrigerated storage ensures longevity while ambient shipping conditions simplify logistics.
Catalog No. NC3640625 Supplier Equitech Bio Inc Supplier No. FSA620100
EQUITECH BIO INC Feline Serum Albumin is a high-purity lyophilized powder ideal for various scientific applications. With ≥ 96% purity and preserved components such as fatty acids, it serves as a versatile serological diluent and molecular weight standard. Processed under cold, non-denaturing conditions, it maintains protein integrity, making it suitable for research and manufacturing purposes. Refrigerated storage ensures longevity while ambient shipping conditions simplify logistics.