Learn More
Med Vet International Feline "V" Notch Elevator, CVD, Outside Grooved, J0899A, Each
Catalog No. 5000585275
Encompass_Preferred
Modified with a "V" cut into the end tip of this instrument, this helps the tip stay on the tooth surface and guides into the socket, preventing it from slipping and damaging the gingiva. The curved tip is designed to follow the root curves of teeth, (i.e., canine and incisor teeth). The elevator tip can be on either the outside or inside of the curved end.
Catalog No. 50-005-85275 Supplier Med Vet International Supplier No. JOR899A
Modified with a "V" cut into the end tip of this instrument, this helps the tip stay on the tooth surface and guides into the socket, preventing it from slipping and damaging the gingiva. The curved tip is designed to follow the root curves of teeth, (i.e., canine and incisor teeth). The elevator tip can be on either the outside or inside of the curved end.