Learn More
Med Vet International Nobivac Feline 2-FELV Vaccine, 25X1 Dose Tray
Catalog No. 502188615
Encompass_Preferred
Feline Leukemia Vaccine, Killed Virus, cats 9 weeks and older. Preservatives: Gentamicin and amphotericin B.Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges*
Catalog No. 50-218-8615 Supplier Med Vet International Supplier No. PHVFELVSP1
Feline Leukemia Vaccine, Killed Virus, cats 9 weeks and older. Preservatives: Gentamicin and amphotericin B.Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges*