Learn More
Selleck Chemical LLC Felodipine S1885-10mg
SDP
Catalog No. 503110742
Encompass_Preferred
Felodipine (CGH-869) is a selective L-type Ca2 channel blocker with IC50 of 0 15 nM
Catalog No. 50-311-0742 Supplier Selleck Chemical LLC Supplier No. S188510MG
Felodipine (CGH-869) is a selective L-type Ca2 channel blocker with IC50 of 0 15 nM