Learn More
Medchemexpress LLC Felodipine 3,5-dimethyl ester | 91189-59-2 | MFCD02673625 | 50mg
SDP
Catalog No. 5000269243
Encompass_Preferred
Felodipine 3 5-Dimethyl Ester is a aryldihydropyridine derivatives for use as mineralocorticoid receptor modulator and voltage-dependent L-type calcium channel CaV1 2 inhibitor[1]
Catalog No. 50-002-69243 Supplier Medchemexpress LLC Supplier No. HYZ776050MG
Felodipine 3 5-Dimethyl Ester is a aryldihydropyridine derivatives for use as mineralocorticoid receptor modulator and voltage-dependent L-type calcium channel CaV1 2 inhibitor[1]