Learn More
Medchemexpress LLC Felodipine | 72509-76-3 | 99.83% | 5 MG
SDP
Catalog No. 5000406668
Encompass_Preferred
Felodipine is a potent, vasoselective dihydropyridine calcium channel antagonist. It lowers blood pressure by selectively acting on vascular smooth muscle, particularly in the resistance vessels. This anti-hypertensive agent also induces autophagy and can cross the blood-brain barrier. It is intended for research use only and not for sale to patients.
  • Potent, vasoselective calcium channel antagonist
  • Lowers blood pressure by acting on vascular smooth muscle
  • Induces autophagy
  • Can cross the blood-brain barrier
Catalog No. 50-004-06668 Supplier Medchemexpress LLC Supplier No. HYB03095MG
Felodipine is a potent, vasoselective dihydropyridine calcium channel antagonist. It lowers blood pressure by selectively acting on vascular smooth muscle, particularly in the resistance vessels. This anti-hypertensive agent also induces autophagy and can cross the blood-brain barrier. It is intended for research use only and not for sale to patients.
  • Potent, vasoselective calcium channel antagonist
  • Lowers blood pressure by acting on vascular smooth muscle
  • Induces autophagy
  • Can cross the blood-brain barrier