Learn More
Medchemexpress LLC Felodipine | 72509-76-3 | 99.83% | 50 MG
SDP
Catalog No. 5000406548
Encompass_Preferred
Felodipine is a dihydropyridine, a potent and vasoselective calcium channel antagonist. It effectively lowers blood pressure by targeting vascular smooth muscle, particularly in resistance vessels. This anti-hypertensive agent also possesses the ability to induce autophagy and can penetrate the blood-brain barrier.
  • Potent vasoselective calcium channel antagonist
  • Lowers blood pressure by acting on vascular smooth muscle
  • Induces autophagy
  • Able to cross the blood-brain barrier
Catalog No. 50-004-06548 Supplier Medchemexpress LLC Supplier No. HYB030950MG
Felodipine is a dihydropyridine, a potent and vasoselective calcium channel antagonist. It effectively lowers blood pressure by targeting vascular smooth muscle, particularly in resistance vessels. This anti-hypertensive agent also possesses the ability to induce autophagy and can penetrate the blood-brain barrier.
  • Potent vasoselective calcium channel antagonist
  • Lowers blood pressure by acting on vascular smooth muscle
  • Induces autophagy
  • Able to cross the blood-brain barrier