Learn More
Selleck Chemical LLC Felodipine 50mg 72509-76-3 CGH-869
SDP
Catalog No. 505410
Encompass_Preferred
Felodipine (CGH-869) is a selective L-type Ca2+ channel blocker with IC50 of 0.15 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-541-0 Supplier Selleck Chemical LLC Supplier No. S188550MG
Felodipine (CGH-869) is a selective L-type Ca2+ channel blocker with IC50 of 0.15 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.