Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Felodipine, Tocris Bioscience™

Catalog No. 296010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-601-0 Supplier R&D Systems Supplier No. 2960/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Ca2+ channel blocker (L-type)

L-type Ca2+ channel blocker that is selective over N-, R-, P/Q- and T-type channels. Displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.

Chemical Identifiers

CAS 72509-76-3
Molecular Formula C18H19Cl2NO4
Molecular Weight (g/mol) 384.253
InChI Key RZTAMFZIAATZDJ-UHFFFAOYSA-N
Synonym felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex
PubChem CID 3333
ChEBI CHEBI:585948
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C

Specifications

Quantity 10 mg
Formula Weight 384.25
Percent Purity >98%
Chemical Name or Material Felodipine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.