Learn More
Sarstedt Inc Felt marker, black, Alcohol-resistant, 10/pack
Catalog No. NC1544798 Shop All Sarstedt Inc. Products
Encompass_Preferred
Black Felt pen especially suitable for writing on e.g. dishes, flasks or plates made of polystyrene.
Catalog No. NC1544798 Supplier Sarstedt Inc Supplier No. 95.950
Black Felt pen especially suitable for writing on e.g. dishes, flasks or plates made of polystyrene.