Learn More
TARGETMOL CHEMICALS INC Felypressin acetate 25MG
SDP
Catalog No. 502877504
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Felypressin acetate (PLV-2) is an agonist of vasopressin 1 and acts on all arginine vasopressin receptors 1AS. Felypressin acetate can be used in dental procedures. Purity 97.06%
Catalog No. 50-287-7504 Supplier TARGETMOL CHEMICALS INC Supplier No. T2568L25MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Felypressin acetate (PLV-2) is an agonist of vasopressin 1 and acts on all arginine vasopressin receptors 1AS. Felypressin acetate can be used in dental procedures. Purity 97.06%