Learn More
New Jersey Fluid Systems LLC FEM BR TEE 1/4T-1/4FPT
Catalog No. NC0398032
Encompass
316 Stainless Steel Swagelok Tube Fitting Female Branch Tee. 1/4 in. OD x 1/4 in. Female NPT
Catalog No. NC0398032 Supplier New Jersey Fluid Systems LLC Supplier No. SS40034TTF
316 Stainless Steel Swagelok Tube Fitting Female Branch Tee. 1/4 in. OD x 1/4 in. Female NPT