Learn More
Aviva Systems Biology Corp FEM1B Antibody - middle region (ARP53702_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53702_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FEM1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FEM1B. Alternate names: DKFZp451E0710, FIAA, F1A-ALPHAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61099