Learn More
World Precision Instrument Female Luer Fitting for 1/16" ID Tubing
SDP
Catalog No. 50822000
Encompass
  • Female Luer Fitting for 1/16" ID Tubing, Polypropylene
  • Package of 100
Catalog No. 50-822-000 Supplier World Precision Instrument Supplier No. 13156100
  • Female Luer Fitting for 1/16" ID Tubing, Polypropylene
  • Package of 100