Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Female Luer to Hose Barb Adapter, 1/4"" ID; 25/PK
Catalog No. NC0227402
Encompass
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
Catalog No. NC0227402 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4550220
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.