Learn More
Vici Valco FEMALE 1/4-28 TO MALE 1/8" N
SDP
Catalog No. NC1752686
Encompass_Preferred
Female 1/4-28 to male 1/8" NPT, CTFE adapter
Catalog No. NC1752686 Supplier Vici Valco Supplier No. CPA2KF
Female 1/4-28 to male 1/8" NPT, CTFE adapter