Learn More
COLDER PRODUCTS COMPANY FEMALE BODY 1/4 ID HB 25/PK
Catalog No. 503131325
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 8 L x 0 94 in H (45 7 x 23 mm) Hose Barb Non-Valved In-line Coupling Body 0 25 in flow coupler x 0 25 in hose barb White For Use With 6 4mm Includes Plastic connector Overall Length 1 8 in 45 7 mm
Catalog No. 50-313-1325 Supplier COLDER PRODUCTS COMPANY Supplier No. PLC17004
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 8 L x 0 94 in H (45 7 x 23 mm) Hose Barb Non-Valved In-line Coupling Body 0 25 in flow coupler x 0 25 in hose barb White For Use With 6 4mm Includes Plastic connector Overall Length 1 8 in 45 7 mm