Learn More
New Jersey Fluid Systems LLC FEMALE CONNECTOR, 1/2 IN. TUBE
Catalog No. NC1937903
Encompass
Stainless Steel Swagelok Tube Fitting, Female Connector, 1/2 in. Tube OD x 1/4 in. Female NPT
Catalog No. NC1937903 Supplier New Jersey Fluid Systems LLC Supplier No. SS81074
Stainless Steel Swagelok Tube Fitting, Female Connector, 1/2 in. Tube OD x 1/4 in. Female NPT