Learn More
New Jersey Fluid Systems LLC FEMALE CONNECTOR (TAPERED THRE
Catalog No. NC9249724
Encompass
Connector, Female; SS Swagelok Tube Fitting; Body Material: SS; Connection 1 Size: 1/4 ft.; Connection 1 Type: Tube Fitting; Connection 2 Size: 1/4 in.; Connection 2 Type: FNPT Taper Thread
Catalog No. NC9249724 Supplier New Jersey Fluid Systems LLC Supplier No. SS40074
Connector, Female; SS Swagelok Tube Fitting; Body Material: SS; Connection 1 Size: 1/4 ft.; Connection 1 Type: Tube Fitting; Connection 2 Size: 1/4 in.; Connection 2 Type: FNPT Taper Thread