Learn More
COLDER PRODUCTS COMPANY FEMALE COUPLING 1/2 250/PK
Catalog No. 503131417
Encompass_Preferred
CPC MPX Series Connectors and Configurations Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple Polycarbonate Overall Length 1 28 in 32 5 mm Hose Barb Non-valved Coupling Body Height 1 96 in 50 mm Maximum Pressure 60 psi
Catalog No. 50-313-1417 Supplier COLDER PRODUCTS COMPANY Supplier No. MPX17803
CPC MPX Series Connectors and Configurations Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple Polycarbonate Overall Length 1 28 in 32 5 mm Hose Barb Non-valved Coupling Body Height 1 96 in 50 mm Maximum Pressure 60 psi