Learn More
COLDER PRODUCTS COMPANY FEMALE COUPLING 1/4 250/CSFEM
Catalog No. 503131438
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-valved Coupling Body Polysulfone Diameter Inner 6 4 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber For Use With 6 4mm Cell culture media packaging and transfer lines harvest and collection applications
Catalog No. 50-313-1438 Supplier COLDER PRODUCTS COMPANY Supplier No. MPCK17004T39
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-valved Coupling Body Polysulfone Diameter Inner 6 4 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber For Use With 6 4mm Cell culture media packaging and transfer lines harvest and collection applications