Learn More
COLDER PRODUCTS COMPANY CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polysulfone Diameter Inner 9 5 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Catalog No. 503131432
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polysulfone Diameter Inner 9 5 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Catalog No. 50-313-1432 Supplier COLDER PRODUCTS COMPANY Supplier No. MPC17006T39
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polysulfone Diameter Inner 9 5 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber