Learn More
COLDER PRODUCTS COMPANY CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polycarbonate Diameter Inner 9 5 mm 0 37 in Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple
Catalog No. 503131431
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polycarbonate Diameter Inner 9 5 mm 0 37 in Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple
This item is not returnable. View return policy
Catalog No. 50-313-1431 Supplier COLDER PRODUCTS COMPANY Supplier No. MPC17006T03
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polycarbonate Diameter Inner 9 5 mm 0 37 in Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple