Learn More
SAI Infusion Technologies 12" Extension Line - PU, with Female Luer, 5/pk
SDP
Catalog No. NC1850996
Encompass
12" PU Extension Line with female luer plus open end compatible with a 27G pin. These extension lines are sterile, 5/pk. Product is for laboratory animal research only.
Catalog No. NC1850996 Supplier SAI Infusion Technologies Supplier No. EXTPF1227
12" PU Extension Line with female luer plus open end compatible with a 27G pin. These extension lines are sterile, 5/pk. Product is for laboratory animal research only.