Learn More
IDEX Health & Science Upchurch FEMALE LER THREAD ADAPTOR
Catalog No. NC0669715
Encompass_Preferred
Our Luer-to-MicroTight Adapter is ideal for infusing sample into lab-on-a-chip devices. It is designed to easily connect 360µm OD tubing to a syringe. This assembly comes with (1) F-152 MicroFerrule and (1) P-416 MicroNut.
Catalog No. NC0669715 Supplier IDEX Health & Science Upchurch Supplier No. P662
Our Luer-to-MicroTight Adapter is ideal for infusing sample into lab-on-a-chip devices. It is designed to easily connect 360µm OD tubing to a syringe. This assembly comes with (1) F-152 MicroFerrule and (1) P-416 MicroNut.